| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300019158.mm.1 |
NM_001085383.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019158.mm.1 |
NM_001085383.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019158.mm.1 |
NM_023628.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019158.mm.1 |
NM_023628.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019158.mm.1 |
TC0300002473.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300019159.mm.1 |
NM_001085383.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019159.mm.1 |
NM_001085383.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019159.mm.1 |
NM_023628.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019159.mm.1 |
NM_023628.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300019159.mm.1 |
TC0300002473.mm.1 |
MTA TC |