Assay Details
| Gene Symbol: Otud7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025613.1 | NP_001020784.1 | 6 - 7 | 1022 | 66 | |
| NM_001025614.1 | NP_001020785.1 | 5 - 6 | 964 | 66 | ||
| GenBank mRNA | AK044701.1 | - | 6 - 7 | 1023 | 66 | |
| BC037040.1 | - | 8 - 9 | 1008 | 66 | ||
| BC141397.1 | - | 5 - 6 | 854 | 9056010 | 66 | |
| BC141398.1 | - | 5 - 6 | 854 | 9056011 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Otud7b
Genomic Map