Assay Details
| Gene Symbol: Otud7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025613.1 | NP_001020784.1 | 10 - 11 | 1540 | 70 | |
| NM_001025614.1 | NP_001020785.1 | 9 - 10 | 1482 | 70 | ||
| GenBank mRNA | AK044701.1 | - | 10 - 11 | 1541 | 70 | |
| BC009124.1 | - | 4 - 5 | 449 | 70 | ||
| BC037040.1 | - | 12 - 13 | 1526 | 70 | ||
| BC141397.1 | - | 9 - 10 | 1372 | 9056010 | 70 | |
| BC141398.1 | - | 9 - 10 | 1372 | 9056011 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Otud7b
Genomic Map