Assay Details
| Gene Symbol: C920021L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040446.1 | 1 - 2 | 160 | 86 | ||
| GenBank mRNA | AK044447.1 | - | 1 - 2 | 157 | 86 | |
| AK160363.1 | - | 1 - 2 | 160 | 86 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC028528 | BC028528 | L259 | Anti-sense | Chr. 3 - 95882352 - 95892949 on NCBI Build GRCm38 | |
| Downstream | anterior pharynx defective 1a homolog (C. elegans) | Aph1a |
6530402N02Rik, APH-1a |
7.0kb | Sense | Chr. 3 - 95893907 - 95898592 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C920021L13Rik
Genomic Map