Assay Details
| Gene Symbol: C920021L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040446.1 | 3 - 4 | 700 | 110 | ||
| GenBank mRNA | AK044447.1 | - | 3 - 4 | 697 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cDNA sequence BC028528 | BC028528 | L259 | Anti-sense | Chr. 3 - 95882352 - 95892949 on NCBI Build GRCm38 | |
| Downstream | anterior pharynx defective 1a homolog (C. elegans) | Aph1a |
6530402N02Rik, APH-1a |
4.8kb | Sense | Chr. 3 - 95893907 - 95898592 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C920021L13Rik
Genomic Map