Assay Details
| Gene Symbol: Aph1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146104.3 | NP_666216.1 | 3 - 4 | 602 | 80 | |
| NM_146134.2 | NP_666246.1 | 3 - 4 | 602 | 80 | ||
| GenBank mRNA | AK078343.1 | - | 1 - 2 | 67 | 80 | |
| AK148400.1 | - | 3 - 4 | 555 | 80 | ||
| AK152334.1 | - | 3 - 4 | 545 | 80 | ||
| AX775953.1 | - | 3 - 4 | 466 | 80 | ||
| BC012406.1 | - | 3 - 4 | 527 | 3483214 | 80 | |
| BC024111.1 | - | 3 - 4 | 468 | 5098973 | 80 | |
| BC057865.1 | - | 3 - 4 | 545 | 5320175 | 80 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Aph1a
Genomic Map