Assay Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039376.2 | NP_001034465.2 | 18 - 19 | 2546 | 108 | |
| NM_001289701.1 | NP_001276630.1 | 18 - 19 | 2546 | 108 | ||
| NM_001289702.1 | NP_001276631.1 | 18 - 19 | 2546 | 108 | ||
| NM_178080.4 | NP_835181.2 | 11 - 12 | 3054 | 108 | ||
| NR_110360.1 | 11 - 12 | 3568 | 108 | |||
| GenBank mRNA | AK028810.1 | - | 11 - 12 | 3021 | 108 | |
| AK122286.1 | - | 11 - 12 | 3049 | 108 | ||
| AK141917.1 | - | 18 - 19 | 2672 | 108 | ||
| BC025653.1 | - | 11 - 12 | 2980 | 108 | ||
| BC028274.1 | - | 18 - 19 | 2556 | 108 | ||
| BC034266.1 | - | 11 - 12 | 1719 | 108 | ||
| BC050783.1 | - | 11 - 12 | 3008 | 108 | ||
| BC057350.1 | - | 11 - 12 | 3485 | 108 | ||
More Information
Set Membership: |
|

Pde4dip
Genomic Map