Assay Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039376.2 | NP_001034465.2 | 20 - 21 | 2723 | 82 | |
| NM_001289701.1 | NP_001276630.1 | 20 - 21 | 2723 | 82 | ||
| NM_001289702.1 | NP_001276631.1 | 20 - 21 | 2723 | 82 | ||
| NM_178080.4 | NP_835181.2 | 13 - 14 | 3231 | 82 | ||
| NR_110360.1 | 13 - 14 | 3745 | 82 | |||
| GenBank mRNA | AK028810.1 | - | 13 - 14 | 3198 | 82 | |
| AK122286.1 | - | 13 - 14 | 3226 | 82 | ||
| BC034266.1 | - | 13 - 14 | 1896 | 82 | ||
| BC050783.1 | - | 13 - 14 | 3185 | 82 | ||
| BC057350.1 | - | 13 - 14 | 3662 | 82 | ||
More Information
Set Membership: |
|

Pde4dip
Genomic Map