Assay Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039376.2 | NP_001034465.2 | 2 - 3 | 270 | 64 | |
| NM_001289701.1 | NP_001276630.1 | 2 - 3 | 270 | 64 | ||
| NM_001289702.1 | NP_001276631.1 | 2 - 3 | 270 | 64 | ||
| GenBank mRNA | AK051209.1 | - | 2 - 3 | 348 | 64 | |
| AK083867.1 | - | 2 - 3 | 345 | 64 | ||
| AK141917.1 | - | 2 - 3 | 396 | 64 | ||
| BC028274.1 | - | 2 - 3 | 280 | 64 | ||
| BC141185.1 | - | 2 - 3 | 270 | 9055797 | 64 | |
| BC145504.1 | - | 2 - 3 | 270 | 9054067 | 64 | |
More Information
Set Membership: |
|

Pde4dip
Genomic Map