Assay Details
| Gene Symbol: Akr1cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027582.3 | NP_081858.2 | 3 - 4 | 451 | 71 | |
| GenBank mRNA | AK014937.1 | - | 2 - 3 | 330 | 71 | |
| AK135492.1 | - | 3 - 4 | 454 | 71 | ||
| BC051128.1 | - | 3 - 4 | 451 | 6707386 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akr1cl
Genomic Map