Assay Details
| Gene Symbol: Fzd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022721.3 | NP_073558.2 | 1 - 2 | 619 | 54 | |
| GenBank mRNA | AK030111.1 | - | 1 - 2 | 621 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030111.1, NM_022721.3 transcripts.
Set Membership: |
|

Fzd5
Genomic Map