Assay Details
| Gene Symbol: Ccnyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097644.1 | NP_001091113.1 | 6 - 7 | 362 | 99 | |
| GenBank mRNA | AK132782.1 | - | 717 | 99 | ||
| CN668580.1 | - | 2 - 3 | 80 | 99 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ccnyl1
Genomic Map