Assay Details
| Gene Symbol: Prkab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182997.2 | NP_892042.2 | 2 - 3 | 337 | 68 | |
| GenBank mRNA | BC060228.1 | - | 2 - 3 | 337 | 6839848 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Prkab2
Genomic Map