Assay Details
| Gene Symbol: Prkab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182997.2 | NP_892042.2 | 6 - 7 | 851 | 80 | |
| GenBank mRNA | AK037124.1 | - | 1 - 2 | 185 | 80 | |
| BC060228.1 | - | 6 - 7 | 851 | 6839848 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkab2
Genomic Map