Assay Details
| Gene Symbol: Psg29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054064.3 | NP_473405.1 | 3 - 4 | 964 | 114 | |
| GenBank mRNA | AK141330.1 | - | 3 - 4 | 965 | 114 | |
| BC114207.1 | - | 3 - 4 | 941 | 30083585 | 114 | |
More Information
Set Membership: |
|

Psg29
Genomic Map