Assay Details
| Gene Symbol: Stat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205313.1 | NP_001192242.1 | 8 - 9 | 930 | 87 | |
| NM_001205314.1 | NP_001192243.1 | 8 - 9 | 966 | 87 | ||
| NM_009283.4 | NP_033309.3 | 8 - 9 | 981 | 87 | ||
| GenBank mRNA | AK018544.1 | - | 8 - 9 | 932 | 87 | |
| AK039458.1 | - | 8 - 9 | 982 | 87 | ||
| AK046517.1 | - | 8 - 9 | 930 | 87 | ||
| AK082706.1 | - | 8 - 9 | 963 | 87 | ||
| AK084855.1 | - | 8 - 9 | 915 | 87 | ||
| AK088874.1 | - | 7 - 8 | 771 | 87 | ||
| AK157263.1 | - | 8 - 9 | 830 | 87 | ||
| AK159887.1 | - | 8 - 9 | 965 | 87 | ||
| BC004808.1 | - | 8 - 9 | 917 | 3587831 | 87 | |
| U06924.1 | - | 6 - 7 | 635 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stat1
Genomic Map