Assay Details
| Gene Symbol: Gls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081081.2 | NP_001074550.1 | 1 - 2 | 636 | 75 | |
| NM_001113383.1 | NP_001106854.1 | 1 - 2 | 636 | 75 | ||
| GenBank mRNA | AK173039.1 | - | 1 - 2 | 636 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gls
Genomic Map