Assay Details
| Gene Symbol: Gls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081081.2 | NP_001074550.1 | 8 - 9 | 1328 | 130 | |
| NM_001113383.1 | NP_001106854.1 | 8 - 9 | 1328 | 130 | ||
| GenBank mRNA | AK046297.1 | - | 3 - 4 | 250 | 130 | |
| AK048072.1 | - | 6 - 7 | 506 | 130 | ||
| AK173039.1 | - | 8 - 9 | 1328 | 130 | ||
| BC025534.1 | - | 2 - 3 | 112 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gls
Genomic Map