Assay Details
| Gene Symbol: Sv2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022030.3 | NP_071313.1 | 9 - 10 | 1974 | 114 | |
| GenBank mRNA | AK028318.1 | - | 9 - 10 | 1919 | 114 | |
| AK080502.1 | - | 9 - 10 | 1928 | 114 | ||
| AK080525.1 | - | 9 - 10 | 2017 | 114 | ||
| AK122360.1 | - | 9 - 10 | 2010 | 114 | ||
| AK144166.1 | - | 9 - 10 | 1997 | 114 | ||
| BC026494.1 | - | 3 - 4 | 321 | 114 | ||
| BC046587.1 | - | 9 - 10 | 1974 | 6493509 | 114 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK158936.1 | - | Chr. - - [-] on Build | 114 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sv2a
Genomic Map