Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
TC0400004047.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001029837.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001029837.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001164049.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001164049.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001164050.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001164050.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001164051.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001164051.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001164052.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_001164052.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_008840.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400033633.mm.1 |
NM_008840.3 |
Refseq |