Assay Details
| Gene Symbol: Apcdd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133237.3 | NP_573500.2 | 3 - 4 | 1110 | 64 | |
| GenBank mRNA | AK156319.1 | - | 3 - 4 | 1111 | 64 | |
| AK172004.1 | - | 1114 | 64 | |||
| AK186282.1 | - | 38 | 64 | |||
| BC141269.1 | - | 4 - 5 | 884 | 9055881 | 64 | |
| BC141270.1 | - | 4 - 5 | 930 | 9055882 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apcdd1
Genomic Map