Assay Details
| Gene Symbol: Prrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028447.3 | NP_082723.2 | 4 - 5 | 827 | 79 | |
| GenBank mRNA | AK143537.1 | - | 4 - 5 | 819 | 79 | |
| BC014817.1 | - | 4 - 5 | 743 | 3709357 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prrc1
Genomic Map