Assay Details
| Gene Symbol: Prrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028447.3 | NP_082723.2 | 6 - 7 | 1095 | 64 | |
| GenBank mRNA | AK004443.1 | - | 6 - 7 | 1095 | 64 | |
| AK143537.1 | - | 6 - 7 | 1087 | 64 | ||
| BC014817.1 | - | 6 - 7 | 1011 | 3709357 | 64 | |
More Information
Set Membership: |
|

Prrc1
Genomic Map