Assay Details
| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007998.6 | NP_032024.2 | 3 - 4 | 413 | 59 | |
| GenBank mRNA | AK004718.2 | - | 3 - 4 | 414 | 59 | |
| AK018738.2 | - | 3 - 4 | 416 | 59 | ||
| AK084843.1 | - | 3 - 4 | 311 | 59 | ||
| AK150685.1 | - | 3 - 4 | 415 | 59 | ||
| AK159652.1 | - | 3 - 4 | 446 | 59 | ||
| BC006746.1 | - | 3 - 4 | 420 | 3256604 | 59 | |
| BC094548.1 | - | 3 - 4 | 386 | 59 | ||
| BC096770.1 | - | 2 - 3 | 247 | 59 | ||
| M59288.1 | - | 2 - 3 | 250 | 59 | ||
| M61215.1 | - | 3 - 4 | 406 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fech
Genomic Map