Assay Details
| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007998.6 | NP_032024.2 | 8 - 9 | 1013 | 60 | |
| GenBank mRNA | AK004718.2 | - | 8 - 9 | 1014 | 60 | |
| AK018738.2 | - | 8 - 9 | 1016 | 60 | ||
| AK084843.1 | - | 8 - 9 | 911 | 60 | ||
| AK150685.1 | - | 8 - 9 | 1015 | 60 | ||
| AK159652.1 | - | 8 - 9 | 1046 | 60 | ||
| BC006746.1 | - | 8 - 9 | 1020 | 3256604 | 60 | |
| BC094548.1 | - | 8 - 9 | 986 | 60 | ||
| BC096770.1 | - | 7 - 8 | 847 | 60 | ||
| M59288.1 | - | 7 - 8 | 850 | 60 | ||
| M61215.1 | - | 8 - 9 | 1000 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fech
Genomic Map