Assay Details
| Gene Symbol: Atp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001488.3 | NP_001001488.2 | 12 - 13 | 1545 | 140 | |
| GenBank mRNA | AK036959.1 | - | 12 - 13 | 1466 | 140 | |
| AK157316.1 | - | 12 - 13 | 1813 | 140 | ||
| AY506548.1 | - | 12 - 13 | 1274 | 140 | ||
| BC117946.2 | - | 12 - 13 | 1272 | 40098035 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp8b1
Genomic Map