Assay Details
| Gene Symbol: Atp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001488.3 | NP_001001488.2 | 16 - 17 | 2148 | 68 | |
| GenBank mRNA | AK036959.1 | - | 16 - 17 | 2070 | 68 | |
| AK157316.1 | - | 16 - 17 | 2416 | 68 | ||
| AY506548.1 | - | 16 - 17 | 1877 | 68 | ||
| BC117946.2 | - | 16 - 17 | 1875 | 40098035 | 68 | |
More Information
Set Membership: |
|

Atp8b1
Genomic Map