Assay Details
| Gene Symbol: Atp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001488.3 | NP_001001488.2 | 4 - 5 | 719 | 67 | |
| GenBank mRNA | AK036959.1 | - | 4 - 5 | 640 | 67 | |
| AK157316.1 | - | 4 - 5 | 987 | 67 | ||
| AY506548.1 | - | 4 - 5 | 448 | 67 | ||
| BC117946.2 | - | 4 - 5 | 446 | 40098035 | 67 | |
More Information
Set Membership: |
|

Atp8b1
Genomic Map