| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_001164167.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_001164167.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_001164168.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_001164168.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_001164169.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_001164169.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_001164170.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_001164170.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_008602.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
NM_008602.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800005982.mm.1 |
TC1800000825.mm.1 |
MTA TC |