Assay Details
| Gene Symbol: Ppp2r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021529.3 | NP_067504.2 | 5 - 6 | 633 | 122 | |
| GenBank mRNA | AB041577.1 | - | 5 - 6 | 622 | 122 | |
| AJ238247.2 | - | 4 - 5 | 460 | 122 | ||
| AK013403.1 | - | 4 - 5 | 429 | 122 | ||
| AK015746.1 | - | 4 - 5 | 462 | 122 | ||
| AK028063.1 | - | 5 - 6 | 598 | 122 | ||
| AK028254.1 | - | 4 - 5 | 552 | 122 | ||
| AK033405.1 | - | 5 - 6 | 632 | 122 | ||
| AK040589.1 | - | 4 - 5 | 461 | 122 | ||
| AK087572.1 | - | 4 - 5 | 456 | 122 | ||
| AK088240.1 | - | 4 - 5 | 542 | 122 | ||
| AK163192.1 | - | 632 | 122 | |||
| AK168298.1 | - | 5 - 6 | 635 | 122 | ||
| BC024754.1 | - | 5 - 6 | 596 | 5375405 | 122 | |
| BC138295.1 | - | 4 - 5 | 429 | 8861314 | 122 | |
| BC138296.1 | - | 4 - 5 | 429 | 8861315 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r3c
Genomic Map