Assay Details
| Gene Symbol: H6pd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291004.1 | NP_001277933.1 | 2 - 3 | 840 | 62 | |
| NM_173371.4 | NP_775547.2 | 2 - 3 | 908 | 62 | ||
| GenBank mRNA | AK045199.1 | - | 2 - 3 | 909 | 62 | |
| AK159373.1 | - | 2 - 3 | 782 | 62 | ||
| AK172249.1 | - | 2 - 3 | 788 | 62 | ||
| BC042677.1 | - | 2 - 3 | 734 | 4195793 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

H6pd
Genomic Map