Assay Details
| Gene Symbol: Clstn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290989.1 | NP_001277918.1 | 14 - 15 | 2464 | 68 | |
| NM_023051.5 | NP_075538.1 | 15 - 16 | 2494 | 68 | ||
| GenBank mRNA | AJ289016.1 | - | 15 - 16 | 2325 | 68 | |
| AK034544.1 | - | 7 - 8 | 1036 | 68 | ||
| AK173059.1 | - | 14 - 15 | 2504 | 68 | ||
| BC029027.1 | - | 10 - 11 | 1579 | 68 | ||
| BC053843.1 | - | 14 - 15 | 2433 | 6315355 | 68 | |
| CX566257.1 | - | 4 - 5 | 564 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Clstn1
Genomic Map