| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400016019.mm.1 |
NM_026963.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016019.mm.1 |
NM_026963.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016019.mm.1 |
TC0400001950.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400016011.mm.1 |
NM_026963.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016011.mm.1 |
NM_026963.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016011.mm.1 |
TC0400001950.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400016013.mm.1 |
NM_026963.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016013.mm.1 |
NM_026963.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016013.mm.1 |
TC0400001950.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400016012.mm.1 |
NM_026963.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016012.mm.1 |
NM_026963.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016012.mm.1 |
TC0400001950.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400016018.mm.1 |
NM_026963.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016018.mm.1 |
NM_026963.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016018.mm.1 |
TC0400001950.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400016010.mm.1 |
NM_026963.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016010.mm.1 |
NM_026963.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016010.mm.1 |
TC0400001950.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400016017.mm.1 |
NM_026963.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016017.mm.1 |
NM_026963.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016017.mm.1 |
TC0400001950.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400016015.mm.1 |
NM_026963.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016015.mm.1 |
NM_026963.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400016015.mm.1 |
TC0400001950.mm.1 |
MTA TC |