Assay Details
| Gene Symbol: Nmnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133435.1 | NP_597679.1 | 3 - 4 | 383 | 59 | |
| GenBank mRNA | AF260925.1 | - | 3 - 4 | 536 | 59 | |
| AK012194.1 | - | 3 - 4 | 617 | 59 | ||
| AK017631.1 | - | 393 | 59 | |||
| AK152407.1 | - | 443 | 59 | |||
| AY679721.1 | - | 3 - 4 | 383 | 59 | ||
| BC138500.1 | - | 3 - 4 | 451 | 8861520 | 59 | |
| BC138502.1 | - | 3 - 4 | 451 | 8861522 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmnat1
Genomic Map