Assay Details
| Gene Symbol: Btf3l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027453.2 | NP_081729.1 | 2 - 3 | 138 | 114 | |
| GenBank mRNA | AK004239.1 | - | 2 - 3 | 169 | 114 | |
| AK013120.1 | - | 2 - 3 | 126 | 114 | ||
| AK076283.1 | - | 2 - 3 | 138 | 114 | ||
| AK167256.1 | - | 2 - 3 | 119 | 114 | ||
| BC024612.1 | - | 2 - 3 | 121 | 114 | ||
| BC024920.1 | - | 2 - 3 | 81 | 114 | ||
| BC058282.1 | - | 2 - 3 | 94 | 30077136 | 114 | |
| BC138215.1 | - | 2 - 3 | 119 | 8861234 | 114 | |
| BC138216.1 | - | 2 - 3 | 119 | 8861235 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004239.1, AK013120.1, AK076283.1, BC058282.1, BC138215.1, BC138216.1, NM_027453.2 transcripts.
Set Membership: |
|

Btf3l4
Genomic Map