Assay Details
| Gene Symbol: Sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047751.1 | - | 1 - 2 | 56 | 57 | |
| AK135543.1 | - | 1 - 2 | 64 | 57 | ||
| AK156828.1 | - | 1 - 2 | 61 | 57 | ||
| BC024418.1 | - | 1 - 2 | 99 | 4982610 | 57 | |
| BC026567.1 | - | 1 - 2 | 87 | 4982239 | 57 | |
| BC085273.1 | - | 1 - 2 | 39 | 30144329 | 57 | |
| CJ060196.1 | - | 1 - 2 | 57 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sf3b4
Genomic Map