Assay Details
| Gene Symbol: Sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047751.1 | - | 3 - 4 | 733 | 62 | |
| AK135543.1 | - | 3 - 4 | 741 | 62 | ||
| AK156828.1 | - | 3 - 4 | 738 | 62 | ||
| BC024418.1 | - | 3 - 4 | 776 | 4982610 | 62 | |
| BC026567.1 | - | 3 - 4 | 764 | 4982239 | 62 | |
| BC085273.1 | - | 3 - 4 | 716 | 30144329 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sf3b4
Genomic Map