Assay Details
| Gene Symbol: Sf3b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047751.1 | - | 5 - 6 | 1114 | 106 | |
| AK135543.1 | - | 5 - 6 | 1122 | 106 | ||
| AK156828.1 | - | 5 - 6 | 1119 | 106 | ||
| BC024418.1 | - | 5 - 6 | 1157 | 4982610 | 106 | |
| BC026567.1 | - | 5 - 6 | 1145 | 4982239 | 106 | |
| BC085273.1 | - | 5 - 6 | 1097 | 30144329 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347939 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347942 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996223 | BC024418.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996223 | BC026567.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996223 | BC085273.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996223 | AK047751.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996223 | AK135543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056293 | AK047751.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2056293 | NM_153053.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | AB221636.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | AF010600.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | AF165911.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | AK141235.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | AK142961.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | AK149897.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | AK166911.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | BC057116.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | BC059240.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | NM_009210.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 554078 | NM_144959.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181656 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147816 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056293 | AK047751.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2056293 | NM_153053.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996223 | BC024418.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996223 | BC026567.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996223 | BC085273.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996223 | AK047751.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996223 | AK135543.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261150 | BC024418.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261150 | BC026567.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261150 | BC085273.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261150 | AK047751.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P261150 | AK135543.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22347942 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147816 | - | - |
| Whole Mouse Genome G4122A | A_52_P253665 | - | - |
| Whole Mouse Genome G4122A | A_51_P261150 | BC024418.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P261150 | BC026567.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P261150 | BC085273.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P261150 | AK047751.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P261150 | AK135543.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101676 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147816 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02762676 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02762678 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03184722 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02762677 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03184721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347939 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347943 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347942 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347938 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22347940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22347876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02488862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347938 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03096187 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02762678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22347955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03184721 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10429453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10429454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494370 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10494371 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424619_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436784_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10494369 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10429452 | - | - |
| Mouse Genome 430 2.0 Array | 1424619_at | - | - |
| Mouse Genome 430 2.0 Array | 1424619_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436784_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1436784_PM_x_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300007419.mm.1 | NM_153053.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300007419.mm.1 | TC0300000926.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436784_x_at | - | - |
| Mouse Expression Set 430 | 1424619_PM_at | - | - |
| Mouse Expression Set 430 | 1424619_at | - | - |
| Mouse Expression Set 430 | 1436784_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424619_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424619_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436784_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436784_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436784_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424619_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436784_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424619_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 110072 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000110072 | - | - |
| mouse-6 v1.1 (Illumina) | scl47090.1_141-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 110072 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47090.1_141-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 110072 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3051392 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3127739 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2612502 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3127739 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3051392 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl47090.1_141-S | - | - |

Sf3b4
Genomic Map