Assay Details
| Gene Symbol: Gpr89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026229.4 | NP_080505.1 | 3 - 4 | 281 | 87 | |
| GenBank mRNA | AK002937.1 | - | 3 - 4 | 301 | 87 | |
| AK016787.1 | - | 3 - 4 | 281 | 87 | ||
| BC010729.1 | - | 3 - 4 | 274 | 3708143 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr89
Genomic Map