Assay Details
Gene Symbol: Zfp236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177832.3 | NP_808500.2 | 2 - 3 | 235 | 119 | |
GenBank mRNA | BC138187.1 | - | 2 - 3 | 235 | 8861206 | 119 |
BC145179.1 | - | 2 - 3 | 235 | 119 | ||
BC145180.1 | - | 2 - 3 | 235 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
