Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800012049.mm.1 |
NM_177832.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012049.mm.1 |
NM_177832.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012049.mm.1 |
TC1800001682.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012044.mm.1 |
NM_177832.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012044.mm.1 |
NM_177832.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012044.mm.1 |
TC1800001682.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012045.mm.1 |
NM_177832.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012045.mm.1 |
NM_177832.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012045.mm.1 |
TC1800001682.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012043.mm.1 |
NM_177832.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012043.mm.1 |
NM_177832.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012043.mm.1 |
TC1800001682.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1800012042.mm.1 |
NM_177832.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012042.mm.1 |
NM_177832.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800012042.mm.1 |
TC1800001682.mm.1 |
MTA TC |