Assay Details
Gene Symbol: Zfp236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177832.3 | NP_808500.2 | 8 - 9 | 1303 | 95 | |
GenBank mRNA | BC138187.1 | - | 8 - 9 | 1303 | 8861206 | 95 |
BC145179.1 | - | 9 - 10 | 1447 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
