Assay Details
| Gene Symbol: Camk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286809.1 | NP_001273738.1 | 1 - 2 | 407 | 62 | |
| NM_177407.4 | NP_803126.1 | 13 - 14 | 1128 | 62 | ||
| GenBank mRNA | AK083245.1 | - | 13 - 14 | 1129 | 62 | |
| AK122410.2 | - | 13 - 14 | 1124 | 62 | ||
| BB659027.1 | - | 1 - 2 | 363 | 62 | ||
| BC031745.1 | - | 13 - 14 | 1109 | 4506005 | 62 | |
| X14836.1 | - | 13 - 14 | 1112 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk2a
Genomic Map