Assay Details
| Gene Symbol: Afg3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027130.1 | NP_081406.1 | 4 - 5 | 529 | 99 | |
| GenBank mRNA | AK009650.1 | - | 4 - 5 | 501 | 99 | |
| AK149970.1 | - | 4 - 5 | 501 | 99 | ||
| AK151207.1 | - | 4 - 5 | 552 | 99 | ||
| AK152115.1 | - | 4 - 5 | 560 | 99 | ||
| AK152727.1 | - | 4 - 5 | 560 | 99 | ||
| AK164537.1 | - | 4 - 5 | 570 | 99 | ||
| BC021380.1 | - | 4 - 5 | 526 | 99 | ||
| BC036999.1 | - | 4 - 5 | 573 | 5042649 | 99 | |
| BC043056.1 | - | 4 - 5 | 529 | 6419521 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afg3l2
Genomic Map