Assay Details
| Gene Symbol: Afg3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027130.1 | NP_081406.1 | 6 - 7 | 756 | 90 | |
| GenBank mRNA | AK080118.1 | - | 1 - 2 | 90 | ||
| AK149970.1 | - | 6 - 7 | 728 | 90 | ||
| AK151207.1 | - | 6 - 7 | 779 | 90 | ||
| AK152115.1 | - | 6 - 7 | 787 | 90 | ||
| AK152727.1 | - | 6 - 7 | 787 | 90 | ||
| AK164537.1 | - | 6 - 7 | 797 | 90 | ||
| BC021380.1 | - | 6 - 7 | 753 | 90 | ||
| BC036999.1 | - | 6 - 7 | 800 | 5042649 | 90 | |
| BC043056.1 | - | 6 - 7 | 756 | 6419521 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afg3l2
Genomic Map