Assay Details
| Gene Symbol: Mad2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009587.1 | - | 7 - 8 | 904 | 80 | |
| AK150216.1 | - | 7 - 8 | 897 | 80 | ||
| AK165627.1 | - | 7 - 8 | 708 | 80 | ||
| BC011282.1 | - | 7 - 8 | 834 | 3484522 | 80 | |
| BC071264.1 | - | 9 - 10 | 834 | 6516319 | 80 | |
| CO043741.1 | - | 1 - 2 | 80 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392616 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392618 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392617 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2402307 | BC071264.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966863 | AK009587.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966863 | AK150216.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966863 | AK165627.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966863 | NM_027985.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720558 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269882 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118290 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966863 | AK009587.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966863 | AK150216.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966863 | AK165627.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966863 | NM_027985.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2402307 | BC071264.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392626 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23392618 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05443671 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118290 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01013306 | - | - |
| Whole Mouse Genome G4122A | A_51_P179987 | AK009587.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179987 | AK150216.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179987 | AK165627.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P179987 | NM_027985.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137508 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122050 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118290 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04010316 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04010313 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392617 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392616 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23392618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23392582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03837992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03694616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03665792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04010316 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03908138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03278957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03908135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03653899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03569667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392642 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03837993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23392593 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10510289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510298 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510288 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510292 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460348_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10510286 | - | - |
| Mouse Genome 430 2.0 Array | 1460348_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1460348_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71890_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71890_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400015714.mm.1 | NM_027985.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400015714.mm.1 | TC0400001925.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400015713.mm.1 | NM_027985.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400015713.mm.1 | TC0400001925.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460348_PM_at | - | - |
| Mouse Expression Set 430 | 1460348_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460348_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460348_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460348_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460348_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1240358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001240358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2707015 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2707015 | - | - |
| mouse-6 v1.1 (Illumina) | scl24707.8.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1240358 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24707.8.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1240358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2707015 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2707015 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24707.8.1_1-S | - | - |

Mad2l2
Genomic Map