Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140734.1 | - | 13 - 14 | 1448 | 122 | |
| AK220466.1 | - | 13 - 14 | 1457 | 122 | ||
| BC021396.1 | - | 12 - 13 | 1487 | 122 | ||
| BC070412.1 | - | 14 - 15 | 1622 | 5705112 | 122 | |
| U96963.1 | - | 12 - 13 | 1460 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map