Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK220466.1 | - | 14 - 15 | 1531 | 111 | |
| BC021396.1 | - | 13 - 14 | 1561 | 111 | ||
| BC070412.1 | - | 15 - 16 | 1696 | 5705112 | 111 | |
| U96963.1 | - | 13 - 14 | 1534 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map