Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156391.1 | - | 3 - 4 | 652 | 58 | |
| AK220466.1 | - | 16 - 17 | 2401 | 58 | ||
| BC070412.1 | - | 17 - 18 | 2566 | 5705112 | 58 | |
| U96963.1 | - | 15 - 16 | 2404 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map