Assay Details
| Gene Symbol: Diap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156391.1 | - | 6 - 7 | 866 | 74 | |
| AK220466.1 | - | 19 - 20 | 2615 | 74 | ||
| BC070412.1 | - | 20 - 21 | 2780 | 5705112 | 74 | |
| U96963.1 | - | 18 - 19 | 2618 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Diap1
Genomic Map